Multiscreen® Filter Plates, Opaque
Proveedor: Merck
MAFCN0B50 MAFBN0B50
MILFMAFCN0B50EA 1780 EUR
MILFMAFCN0B50 736-0179
Multiscreen® Filter Plates, Opaque
Microplacas Placas de filtro
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated or particular laden samples. They are also widely used for clarification of aqueous solutions.
  • Multiple configurations and applications
  • Chemical compatibility
  • Automation-compatible
  • Superior vacuum filtration and filtrate collection
  • Hydrophilic lid
  • Colour code: Opaque
Glass fiber filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration,More Product Information