Waste Disposal Bags, Liquid-Tight Welded
Proveedor: HAMILTON ROBOTICS
199203 199202
736-0844EA 26 EUR
736-0844 736-0843
Waste Disposal Bags, Liquid-Tight Welded
Bolsas Bolsas de eliminación de residuos
These waste bags are ideal for the containment and disposal of used consumables.
Highly resistant, made of 100 µm thick polyethylene
  • Made of 100 µm thick polyethylene that ensures a high resistance to tear and puncturing