AutoLys tubes and racks
Proveedor: HAMILTON ROBOTICS
235459 235478
736-0848EA 247 EUR
736-0848 736-0849
AutoLys tubes and racks
Tubos Tubos robóticos
The AutoLys tube from Hamilton Robotics is a tube-in-tube device designed to automate lysis steps for forensic, genomic or clinical research, food testing and other laboratory applications. Preparation of cleared lysates from solid matrices is a major bottleneck in many laboratories. The AutoLys filter tube is designed to be an automation-friendly solution specific to the Hamilton STAR Line of robots with the AutoLys channel. However, the tubes can also be used for manual lysis processing on the bench top. The AutoLys tube is available in fully tested Biological purityPLUS grade,More Product Information