Eppendorf® Assay/Reader Microplates
Proveedor: EPPENDORF
0030601.572 0030601.670 0030601.807 0030601.904 0030621.670 0030621.905 0030601.475 0030601.700
737-0169EA 347 EUR
737-0169 737-0170 737-0171 737-0172 737-0227 737-0228 737-0248 737-0249
Eppendorf® Assay/Reader Microplates
Microplacas Microplacas HTS
Eppendorf Assay/Reader plates are designed for diverse microplate assays, optimising absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximising reflection
  • Solid black and white assay plates are made of