Deepwell Plates 96-well, 1000 µl
Proveedor: EPPENDORF
0030501.233 0030501.241 0030502.205 0030502.213 0030502.230 0030502.248 0030501.209 0030501.217 0030505.204 0030506.200 0030113.535
737-2508EA 176 EUR
737-2508 737-2509 737-2520 737-2521 737-2523 737-2524 737-2505 737-2506 737-0201 737-0204 737-0251
Deepwell Plates 96-well, 1000 µl
Microplacas Placas Deep Well
Eppendorf Deepwell plates 96-well, 1000 µl offer efficient sample handling with features like OptiTrack® matrix for rapid identification and g-Safe® technology for stability during centrifugation. SafeCode variants provide secure three-level coding, including error correction for reliable identification, ensuring flexibility and accuracy in experimental workflows.
  • OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimised well geometry for maximum sample