Filter plates, 24-well, AcroPrep™
Proveedor: Cytiva (Formerly Pall Lab)
97026 97016 97017 97027 97047 97048 97049 97050 97051 97052 97053 97054 97055 97056 97057 97058 97059 97060 97061 97062 97029 97030 97031 97032 97033 97034 97035 97036
738-0219EA 586 EUR
738-0219 738-0220 738-0222 738-0221 738-0238 738-0237 738-0236 738-0235 738-0234 738-0233 738-0232 738-0231 738-0230 738-0229 738-0228 738-0227 738-0226 738-0225 738-0224 738-0223 738-0246 738-0245 738-0244 738-0243 738-0242 738-0241 738-0240 738-0239
Filter plates, 24-well, AcroPrep™
Microplacas Placas de filtro
The Acroprep 24-well filter plate utilises Pall’s proprietary high performance multi-layer filter media and membranes to offer time savings, strong performance claims and streamlined workflow improvements in a 24-well plate format.
  • Intrinsic Acroprep plate and membrane properties minimise sample loss from non specific binding
  • Dual-layer clarification plate contains a depth filter plus 0,65/0,2 μm Supor EKV PES membrane, and allows clarification of large cellular debris prior to filtration through the 0,65/0,2 μm EKV Supor membrane
  • Dual-layer clarification