(R)-(-)-Linalol, Supelco®

Proveedor: Merck


M-Clarity™ quality level = MQ100

74856-1ML-F 74856-5ML-F
74856-1ML-FEA 99.6 EUR
74856-1ML-F 74856-5ML-F
(R)-(-)-Linalol, Supelco®
(R)-(-)-Linalol
Formula: C₁₀H₁₈O
Número MDL: MFCD00135469
Núm. CAS: 126-91-0
EINECS: 204-811-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82ce51d5bc511db6d03d4e68af905a8066d98ebc&vid=e3db0e57196f76dead014399a5dc4a45bc6ce1cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR