Anti-TRB Armenian Hamster Monoclonal Antibody (violetFluor® 450) [clone: H57-597]
Proveedor: Tonbo Biosciences
75-5961-U100 75-5961-U025
75-5961-U100EA 179 EUR
75-5961-U100 75-5961-U025
Anti-TRB Armenian Hamster Monoclonal Antibody (violetFluor® 450) [clone: H57-597]
Anticuerpos
The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma/delta protein chains, known as the g/d TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactionsMore Product Information