Broad range hydrometers
Proveedor: Alla France
3000TB100/20QP 3050TB065/20QP 3050FG100/20QP
765-0052EA 20.6 EUR
765-0052 765-0053 ALLA3050FG100/20QP
Broad range hydrometers
Hidrómetros
These broad range hydrometers are calibrated at 20 °C.
Mostly dedicated to applications with citric and tartaric acids.