Sodio hidróxido 50% aqueous solution, AR®, Macron Fine Chemicals™

Proveedor: Avantor Performance Materials
Peligro

Sinónimos: Caustic soda

7706-68EA 0 EUR
7706-68
Sodio hidróxido 50% aqueous solution, AR®, Macron Fine Chemicals™
Sodio hidróxido

Precaución: Protect from air to avoid absorption of carbon dioxide.

Formula: NaOH
MW: 40 g/mol
Pto. Ebullición: 1390
Pto. de fusión: 323
Densidad: 1.53 g/cm³
Número MDL: MFCD00003548
Núm. CAS: 1310-73-2
UN: 1824
ADR: 8 II
Índice Merck: 14,08627

Order Now

Resultados test especificaciones

For Laboratory,Research,or Manufacturing Use 49.5 - 52.0 %
Protect from air to avoid absorption of carbon dioxide. Passes Test
Assay (NaOH) 49.5 - 52.0 %
Identification Passes Test
Ammonium Hydroxide Precipitate ≤0.010 %
Chloride (Cl) ≤0.0025 %
Heavy Metals (as Ag) ≤0.0015 %
Iron (Fe) ≤0.0005 %
Nitrogen Compounds (as N) ≤0.0005 %
Potassium (K) ≤0.025 %
Phosphate (PO₄) ≤0.0005 %
Sulfate (SO₄) ≤0.0025 %
Sodium Carbonate (Na₂CO₃) ≤0.1 %
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR