Genomic DNA isolation and purification kits, OmniTemplate™
Proveedor: G Biosciences
786-013
786-013EA 426 EUR
786-013
Genomic DNA isolation and purification kits, OmniTemplate™
Reactivos de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
OmniTemplate™ is specifically designed for the rapid isolation of a DNA template for PCR analysis from mammalian tissue samples, blood and cell cultures. This single tube method produces a high concentration, ready-to-use supply of DNA template for microanalysis, genotyping and a multitude of other applications.
  • Single tube genomic DNA isolation
  • Compatible with PCR and other downstream applications
  • Suitable for blood, cells, and animal tissue samples
  • No toxic chemicals, no phenol, no hazardous waste