Adenosylhomocysteine nucleosidase
Proveedor: G Biosciences
786-427
786-427EA 696 EUR
786-427
Adenosylhomocysteine nucleosidase
Enzimas
This recombinant adenosylhomocysteine nucleosidase (10 µM) converts S-adenosylhomocysteine to S-(deoxy-D-ribos-5-yl)-L-homocysteine and adenine.