2,4,6-Trifenilpirilio tetrafluoroborato, Sigma-Aldrich®

Proveedor: Merck


M-Clarity™ quality level = MQ200

8.18238.0005
8.18238.0005EA 87.6 EUR
8.18238.0005
2,4,6-Trifenilpirilio tetrafluoroborato, Sigma-Aldrich®
2,4,6-Trifenilpirilio tetrafluoroborato
Formula: C₂₃H₁₇BF₄O
Número MDL: MFCD00012001
Núm. CAS: 448-61-3
EINECS: 207-186-4

Order Now

Resultados test especificaciones

Melting Point 252 - 255 °C
Molar Mass 396.2 g/mol
Hill Formula C23H17BF4O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR