Etanol absoluto ≥99.5%, EMPLURA®, Supelco®

Proveedor: Merck
Peligro

Sinónimos: Ethyl alcohol

M-Clarity™ quality level = MQ200

8.18760.1000 8.18760.2500 8.18760.1000_T 8.18760.2500_T 8.18760.9180 8.18760.9025
8.18760.1000EA 28.8 EUR
8.18760.1000 8.18760.2500 MERC8.18760.1000_T MERC8.18760.2500_T 8.18760.9180 8.18760.9025
Etanol absoluto ≥99.5%, EMPLURA®, Supelco®
Etanol

Bio-Based Ethanol
Instead of synthetic ethanol, we use bioethanol produced from grain or sugar cane. High quality, affordability, and ready availability make our bioethanol an obvious choice for a sustainable future.


  • Produced from grain or sugar cane, a renewable source
  • Less toxic than synthetic ethanol (no toxic by-products)
  • Reliable availability
  • Production method is safer for the environment


Certificaciones: M-Clarity™ quality level= MQ200

Formula: C₂H₅OH
Pto. Ebullición: 78,3 °C (1013 hPa)
Pto. de fusión: –117 °C
Densidad: 0,7895 g/cm³ (20 °C)
Pto. de inflamación: 12 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003568
Núm. CAS: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
IR: 01-2119457610-43
Índice Merck: 12,03806

Order Now

Resultados test especificaciones

Purity (GC) ≥ 99.5 %
Identity (IR) conforms
Density (d 20 °C/ 4 °C) 0.789 - 0.790
Evaporation residue ≤ 0.0025 %
Water ≤ 0.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR