Anti-CD45R Rat Antibody (redFluor™ 710) [clone: RA3-6B2]
Proveedor: Tonbo Biosciences
80-0452-U100 80-0452-U025
80-0452-U100EA 136 EUR
80-0452-U100 80-0452-U025
Anti-CD45R Rat Antibody (redFluor™ 710) [clone: RA3-6B2]
Anticuerpos
The RA3-6B2 antibody reacts with the human and mouse CD45 isoform known as CD45R, or B220, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns, all critical for leukocyte function. In mouse, the CD45R/B220 isoform is predominantly found on B cells, at varying levels on all stages from pro-B cells to activated B cells, and may also be detected on certain T cell and NK cell subsets. It is of note that B220More Product Information