Isoelectric focusing system, Ettan™ IPGphor™ 3
Proveedor: Cytiva
80-6416-11 80-6416-49 80-6416-68 80-6416-87 80-6417-25 80-6417-44 80-6469-88 80-6470-07 80-6455-44 80-6498-38 80-6498-57 28-9334-92 80-6416-30 11-0025-80 28-9384-45 28-9334-65 11-0026-88
80-6416-11EA 3010 EUR
80-6416-11 80-6416-49 80-6416-68 80-6416-87 80-6417-25 80-6417-44 80-6469-88 80-6470-07 80-6455-44 80-6498-38 80-6498-57 28-9334-92 80-6416-30 11-0025-80 28-9384-45 28-9334-65 11-0026-88
Isoelectric focusing system, Ettan™ IPGphor™ 3
Sistemas de electroforesis Sistemas horizontales de electroforesis en gel
Ettan™ IPGphor™ 3 is a fully integrated isoelectric focusing (IEF) system optimised to deliver high throughput, speed, reproducibility, and high protein-loading capacity. Ettan™ IPGphor™ 3 is optimised for easy handling of CyDye™ labelled proteins and other light-sensitive samples, and provides temperature control critical for reproducibility in 2-D DIGE experiments.
  • Temperature control from 15 to 31 °C
  • Integrated high-voltage DC power supply delivering up to 10 kV makes the unit space-saving
  • Secure light protection during the run - compatible with CyDye™-