Poly(vinyl chloride), carboxylated 1,8% carboxyl basis, Selectophore™ ionophore, Supelco®

Proveedor: Merck
Atención


M-Clarity™ quality level = MQ100

81395-50G
81395-50GEA 263 EUR
81395-50G
Poly(vinyl chloride), carboxylated 1,8% carboxyl basis, Selectophore™ ionophore, Supelco®
Poly(vinyl chloride), carboxylated

Poly(vinyl chloride) carboxylated may be used in the preparation of ultrafiltration membranes using phase-inversion method.

Formula: (C₅H₇ClO₂)n
Número MDL: MFCD00084460
Núm. CAS: 25702-80-1
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74f1e6aa0ab080fe44817667892ba44ec27bfb41&vid=7a360a48ddd14f08dba58194d54155dd36f522a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR