Cleanroom labels, gloss
Proveedor: Texwipe
TX542G TX532G TX515G
817-0307EA 859 EUR
817-0307 817-0304 817-0300
Cleanroom labels, gloss
Etiquetas
Glossy cleanroom labels are designed for thermal transfer printing in a cleanroom environment. Available with two different adhesive levels to provide excellent adhesion to a variety of surfaces without leaving residue.
  • Removable adhesive
  • Ideal for applications where ink adhesion and reflectivity are desired