Tough-Tags® label rolls
Proveedor: DIVERSIFIED BIOTECH
TTLC-1000 TTLW-1000 TTSC-1000 TTSW-1000 TTSB-1000 TTSY-1000 TTSR-1000 TTSG-1000 TT-TNY TT-SWALL TTLR-1000 TTLY-1000 TTLG-1000 TTLB-1000 TTGP-1000 TTLO-1000 TTSO-1000
817-2111EA 133 EUR
817-2111 817-2112 817-2113 817-2114 817-2115 817-2116 817-2129 817-2128 817-2130 817-0052 817-2132 817-2133 817-2135 817-2134 DIVETTGP-1000 DIVETTLO-1000 DIVETTSO-1000
Tough-Tags® label rolls
Etiquetas
These Tough-Tags® polyester label rolls are pre-cut, peel-off labels which accept many marking instruments. They are perfectly sized to fit microcentrifuge tubes and other containers in the laboratory.
  • Temperature resistant from −40 to +121 °C
  • Chemically inert, resist most organic solvents and caustic agents
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
  • Can be written on with pen, pencil or permanent marker
Tough-Tags® resist boiling water baths, autoclaving and moderate freezer temperatures. Tubes labeled with Tough-More Product Information