Cleanroom bond paper, TexBond® 22
Proveedor: Texwipe
TX5611 TX5619 TX5623 TX5624 TX5628 TX5629
818-0262EA 30.1 EUR
818-0262 818-0263 818-0264 818-0265 818-0266 818-0267
Cleanroom bond paper, TexBond® 22
Papeles
Medium-weight cellulose paper saturated with synthetic latex substantially reduces particle generation associated with standard papers. These papers are available in DIN A4 and A3 sizes. Accepts writing and printing/copying on all common office equipment.
  • Excellent retention of inks and toners
  • Low particle and ion release
  • Latex-impregnated for optimal purity and smoothness
  • Minimal static charging prevents pages from sticking
  • Sterilisable
  • Weight: 75 g/m²