Ácido nítrico ≥69,0% (by titrimetric analysis), TraceSELECT™ para análisis de trazas, Fluka™

Proveedor: Honeywell Chemicals
Peligro

84385-500ML 84385-6X500ML 84385-1L 84385-2.5L 84385-5L
84385-500MLEA 109 EUR
84385-500ML 84385-6X500ML 84385-1L 84385-2.5L 84385-5L
Ácido nítrico ≥69,0% (by titrimetric analysis), TraceSELECT™ para análisis de trazas, Fluka™
Ácido nítrico

TraceSELECT™ Ultra and TraceSELECT


  • Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

Formula: HNO₃
Pto. Ebullición: 122 °C (1,013 hPa)
Pto. de fusión: –38 °C
Densidad: 1,420 g/cm3 (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011349
Núm. CAS: 7697-37-2
UN: 2031
ADR: 8,II
Índice Merck: 13,06608

Order Now

Resultados test especificaciones

Assay 67 - 70 %
Silver (Ag) Max. 0.1 ppb
Aluminium (Al) Max. 1 ppb
Arsenic (As) Max. 0.5 ppb
Gold (Au) Max. 0.1 ppb
Boron (B) Max. 1 ppb
Barium (Ba) Max. 0.1 ppb
Beryllium (Be) Max. 0.1 ppb
Bismuth (Bi) Max. 0.1 ppb
Calcium (Ca) Max. 1 ppb
Cadmium (Cd) Max. 0.5 ppb
Cerium (Ce) Max. 0.1 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.1 ppb
Copper (Cu) Max. 0.5 ppb
Dysprosium (Dy) Max. 0.1 ppb
Erbium (Er) Max. 0.1 ppb
Europium (Eu) Max. 0.1 ppb
Iron (Fe) Max. 1 ppb
Gallium (Ga) Max. 0.1 ppb
Gadolinium (Gd) Max. 0.1 ppb
Germanium (Ge) Max. 0.1 ppb
Hafnium (Hf) Max. 0.1 ppb
Mercury (Hg) Max. 0.1 ppb
Holmium (Ho) Max. 0.1 ppb
Indium (In) Max. 0.1 ppb
Potassium (K) Max. 1 ppb
Lanthanum (La) Max. 0.1 ppb
Lithium (Li) Max. 0.1 ppb
Lutetium (Lu) Max. 0.1 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 0.1 ppb
Molybdenum (Mo) Max. 0.1 ppb
Sodium (Na) Max. 1 ppb
Niobium (nb) Max. 0.1 ppb
Neodymium (nd) Max. 0.1 ppb
Nickel (Ni) Max. 0.5 ppb
Lead (Pb) Max. 0.1 ppb
Palladium (Pd) Max. 0.5 ppb
Praseodymium (Pr) Max. 0.1 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.1 ppb
Rhenium (Re) Max. 0.1 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Scandium (Sc) Max. 0.1 ppb
Selenium (Se) Max. 1 ppb
Samarium (Sm) Max. 0.1 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.1 ppb
Tantalum (Ta) effective ppb
Terbium (Tb) Max. 0.1 ppb
Tellurium (Te) Max. 0.1 ppb
Thorium (Th) Max. 0.1 ppb
Titanium (Ti) Max. 0.5 ppb
Thallium (Tl) Max. 0.1 ppb
Thulium (Tm) Max. 0.1 ppb
Uran (U) Max. 0.1 ppb
Vanadium (V) Max. 0.5 ppb
Tungsten (W) Max. 0.1 ppb
Yttrium (Y) Max. 0.1 ppb
Ytterbium (Yb) Max. 0.1 ppb
Zinc (Zn) Max. 0.5 ppb
Zirconium (Zr) Max. 0.1 ppb
Chloride (Cl) Max. 0.2 ppm
Phosphate (PO₄) Max. 0.01 ppm
Sulfate (SO₄) Max. 0.3 ppm
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR