VWR® Media Supplements for Dehydrated Culture Media
Proveedor: VWR Chemicals
992830ZFEA 16.4 EUR
992830ZF 301950ZK 84729.0001 84730.0001 84732.0001 84722.0001 84734.0001 84742.0001 84743.0001 84744.0001 84745.0001 84746.0001 84733.0001 84725.0001 84726.0001 84727.0001 84728.0001 84731.0001 84724.0001 84723.0001 84720.0001 84721.0001 84747.0001 928580NL 928610NL 900010ZK 85861.0001 85860.0001 928570NL 85886.0001 85884.0001 928880NL
VWR® Media Supplements for Dehydrated Culture Media
Medios Aditivos y suplementos para microbiología
The isolation of microorganisms often requires the use of specific supplements. The supplements presented here are available in vials and bottles, and the corresponding culture medium to use with them is indicated in the table below. Usually each vial is used to supplement 500 ml of media unless otherwise stated (complete information available on the specification sheets available on request).
  • Freeze-dried