VWR®, UV/Visible standards
Proveedor: VWR Chemicals
86124.001EA 244 EUR
86124.001 86125.001 86126.001 86127.001 86128.001 86129.001 86130.001
VWR®, UV/Visible standards
Normativas Patrones de espectroscopia
For spectrophotometer calibration according to European Pharmacopoeia (Ph. Eur.)
  • Absorbance control
  • Testing of resolution power
  • Stray light testing
  • Wavelength accuracy testing
In the pharmaceutical industry, a regular check of UV/Visible spectrophotometers according to Ph. Eur. is mandatory. Even in other industries, researchers who are using UV/Visible spectrophotometers, and who are working according to e.g. GMP, GLP, USP or ISO guidelines, need to check and qualify their instruments on a regular basis.