Alizarin red S (sodium salt), AnalaR NORMAPUR® Reag. Ph. Eur. para análisis (C.I. 58005)

Proveedor: VWR Chemicals

Sinónimos: C.I. 58005

86156.150EA 69.6 EUR
86156.150 86156.180
Alizarin red S (sodium salt), AnalaR NORMAPUR® Reag. Ph. Eur. para análisis (C.I. 58005)
Alizarin red S (sodium salt)
Formula: C₁₄H₇NaO₇S·1H₂O
Número MDL: MFCD00013049
Núm. CAS: 130-22-3

Order Now

Resultados test especificaciones

pH Range (Yellow to Purple-Red) 3.7 - 5.2
Identity (IR-Spectrum) Passes Test
Insoluble in Water Passes Test
Absorption Maximum λ1 (NaOH 0.1 M) 592 - 596
Absorption Maximum λ2 (NaOH 0.1 M) 553 - 558
Absorptivity (A1%/1 cm; λ max. 1, NaOH 0.1 M) 305 - 480
Absorptivity (A1%/1 cm; λ max. 2, NaOH 0.1 M) 330 - 520
Related Substances (TLC) Passes Test
Loss on Drying (135 °C) ≤5.0
Suitability for Microscopy Passes Test
Sensitivity to Aluminium Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR