5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) 98 TLC_ASSAY_METHOD dye_Amine-reactive fluorescent

Proveedor: Biotium

Sinónimos: TAMRA SE, 5 isomer, 5-TAMRA, SE, 5-TAMRA-N-succinimidyl ester, 5-Carboxytetramethylrhodamine N-succinimidyl ester

90034
90034.EA 325 EUR
90034.
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) 98 TLC_ASSAY_METHOD dye_Amine-reactive fluorescent
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE)

Full name: 5-Carboxytetramethylrhodamine succinimidyl ester (single isomer). Amine-reactive form of carboxytetramethylrhodamine (TAMRA) single isomer.


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solid soluble in DMSO or DMF
  • Store at -20°C and protect from light

Formula: C₂₉H₂₅N₃O₇
MW: 527.52 g/mol
Temperatura de almacenaje: Congelador
Núm. CAS: 150810-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f9bea755bbfe9d3a53593fb23b1edfb56bcb5b0&vid=6f9bea755bbfe9d3a53593fb23b1edfb56bcb5b0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR