Organic reference standard, 4-Deoxy-D-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
Proveedor: Merck
90061-50MG
90061-50MGEA 487 EUR
90061-50MG
Organic reference standard, 4-Deoxy-D-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
Normativas Estándares orgánicos
4-Deoxyerythronic acid has been associated with uremia and diabetes mellitus type 1, but is also a normal organic acid present in human biofluids, derived presumably from L-threonine.
Order Now
ESPECIFICACIONES
Descripción 4-Deoxy-D-erythronic acid sodium salt hydrate
Formula C₄H₈O₄ ·xNa+ ·yH₂O
Temperatura de almacenamiento 2...8 °C