VWR® Hitachi Chromaster Plus HPLC System
Proveedor: VWR Collection
903-0528EA 13020 EUR
903-0528 903-0525 903-0524 903-0520 903-0501 903-0500 903-0537 903-0554 903-0556 903-0555 903-0557 903-0597 903-0320 903-0367 903-0337 903-0342 903-0343 903-0396 903-0361 903-0345 903-0344 903-0355 903-0358 903-0359 903-0370 903-0371 903-0372 903-0164
VWR® Hitachi Chromaster Plus HPLC System
Equipos cromatografía Sistemas completos para HPLC
Experience the next level of high-performance liquid chromatography with the Chromaster Plus HPLC system. This latest version boasts a sleek new design and enhanced performance features, including the upgraded CM5310 column oven and CM5410 UV detector. The new design offers a modern and user-friendly interface, while the enhanced performance of the CM5310 column oven and CM5410 UV detector ensures superior results. Additionally, users can enjoy peace of mind with an extended warranty coverage.
  • Highly flexible, with a wide range of modules (pumps, detectors); easily adaptable to any