2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®

Proveedor: Avantor
Peligro

Sinónimos: Alcohol isopropílico, IPA

9037-01EA 82.06 EUR
9037-01 9037-03 9037-07
2-Propanol ≥99.0% USP, Multi-Compendial, J.T.Baker®
2-Propanol
Formula: (CH₃)₂CHOH
Pto. Ebullición: 82 °C (1013 hPa)
Pto. de fusión: –89 °C
Densidad: 0,786 g/cm³ (20 °C)
Pto. de inflamación: 12 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011674
Núm. CAS: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
IR: 01-2119457558-25
Índice Merck: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Meets B.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.783 - 0.787
USP - Refractive Index, n²ºD 1.376 - 1.378
USP - Acidity Passes Test
USP - Nonvolatile Residue ≤0.005 %
USP - Assay ≥99.0 %
USP - Water (H₂O) ≤0.5 %
USP - Volatile Impurities - Ethyl ether ≤0.1 %
USP - Volatile Impurities - Acetone ≤0.1 %
USP - Volatile Impurities - Diisopropyl ether ≤0.1 %
USP - Volatile Impurities - 1-Propanol ≤0.1 %
USP - Volatile Impurities - 2-Butanol ≤0.1 %
USP - Volatile Impurities - Total ≤1.0 %
USP - Benzene ≤2 ppm
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
Appearance Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Benzene (C₆H₆) ≤2 ppm
EP/BP - Any peak other than 2-butanol ≤0.3 %
EP/BP - Peroxide Passes Test
EP/BP - Nonvolatile Matter ≤20 ppm
EP/BP - Water (H₂O) ≤0.5 %
EP/BP - UV Absorbance - 310 nm ≤0.01
EP/BP - UV Absorbance - 290 nm ≤0.02
EP/BP - UV Absorbance - 270 nm ≤0.03
EP/BP - UV Absorbance - 250 nm ≤0.10
EP/BP - UV Absorbance - 230 nm ≤0.30
EP/BP - Spectrum Appearance Passes Test
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Specific Gravity at 20°/20°C 0.785 - 0.788
JP - Clarity of Solution Passes Test
JP - Acidity Passes Test
JP - Residue on Evaporation, mg/20mL ≤1.0
JP - Water (H₂O) ≤0.75 %
JP - Distilling Range(Lower) 81 - 83 °C
JP - Distilling Range (Upper) 81 - 83 °C
Residual solvents – Only the Class 2 solvent cyclohexane and Class 3
solvents are likely to be present. Cyclohexane is controlled below the
USP 467 Option 1 limit. All Class 3 solvents are less than 0.5%.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR