Anti-CD3,CD11b/c,CD19,CD45R/B220,Ly-6G (Gr-1),TER-119 Monoclonal Antibody (redFluor®710, violetFluor®450,PerCP-Cy5.5®,PE-Cy7®,FITC) [clone:53-6.7,M1/70,N418,RA3-6B2,RB6-8C5,M5/114.15.2]

Proveedor: Tonbo Biosciences
91-0003-U025
91-0003-U025EA 386 EUR
91-0003-U025
Anti-CD3,CD11b/c,CD19,CD45R/B220,Ly-6G (Gr-1),TER-119 Monoclonal Antibody (redFluor®710, violetFluor®450,PerCP-Cy5.5®,PE-Cy7®,FITC) [clone:53-6.7,M1/70,N418,RA3-6B2,RB6-8C5,M5/114.15.2]
Anticuerpos
The Mouse Myeloid Precursor backbone panel is a five-color combination of seven anti-mouse antibodies. This panel can serve as a backbone to identify various myeloid precursor populations, and leaves room for GFP or additional drop-in markers, if desired. The CD3e, CD19 and CD45R/B220 positive cells are gated together to exclude mature T and B cells. Violet laser (405 nm) required for detection of CD11b violetFluorTM 450. Hosts: Rat, Armenian Hamster

Type: Primary
Antigen: CD3, CD11b, CD11c, CD19, CD45R/B220, Ly-6G (Gr-1), TER-119
Clonality: Monoclonal
Clone: 53-6.7, M1/70, N418, RA3-6B2, RB6More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR