Amonio nitrato 50 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS

Proveedor: VWR Chemicals

91277.154EA 161 EUR
91277.154
Amonio nitrato 50 g/L (99,999%) in water, ARISTAR® matrix modifier solution, for graphite furnace AAS
Amonio nitrato

The VWR® Matrix modifier solutions have especially been developed for use in GF-AAS to eliminate matrix effects. They can be used either diluted or as mixtures.


  • Guaranteed accuracy: ±1%
  • COA incl. up to 70 measured trace impurities in ppm range


VWR® Matrix modifier solutions are produced with high purity metals or salts (≥99,999%) in sub-boiling distilled acids and are supplied with a detailed batch specific certificate of analysis.

Formula: NH₄NO₃
MW: 80.04 g/mol
Pto. Ebullición: 230
Pto. de fusión: 169
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011425
Núm. CAS: 6484-52-2
Índice Merck: 13,00538

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR