Ácido nítrico ≥70%, CMOS for microelectronic, J.T.Baker®

Proveedor: Avantor
Peligro

Sinónimos: Nitric acid solution

9606-03EA 195 EUR
9606-03
Ácido nítrico ≥70%, CMOS for microelectronic, J.T.Baker®
Ácido nítrico
Formula: HNO₃
MW: 63.01 g/mol
Pto. Ebullición: 87
Pto. de fusión: -42
Densidad: 1.51
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011349
Núm. CAS: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 I
Índice Merck: 13,06610

Order Now

Resultados test especificaciones

Assay (HNO₃} 69.0 - 70.0 %
Appearance Passes Test Passes Test
Color (APHA) ≤10
Residue after Ignition ≤2 ppm
Chloride (Cl) ≤0.08 ppm
Phosphate (PO₄) ≤0.1 ppm
Sulfate (SO₄) ≤0.2 ppm
Trace Impurities - Aluminum (Al) ≤40.0 ppb
Arsenic and Antimony (as As) ≤5 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤20.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤50.0 ppb
Trace Impurities - Calcium (Ca) ≤50.0 ppb
Trace Impurities - Chromium (Cr) ≤30.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤20.0 ppb
Trace Impurities - Gold (Au) ≤20.0 ppb
Heavy Metals (as Pb) ≤100 ppb
Trace Impurities - Iron (Fe) ≤40.0 ppb
Trace Impurities - Lead (Pb) ≤20.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤20.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Nickel (Ni)
Trace Impurities - Niobium (Nb)
Trace Impurities - Potassium (K) ≤50.0 ppb
Trace Impurities - Silicon (Si) ≤50.0 ppb
Trace Impurities - Silver (Ag) ≤20.0 ppb
Trace Impurities - Sodium (Na) ≤150.0 ppb
Trace Impurities - Strontium (Sr) ≤30.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤20.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤20.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count - 0.5 µm and greater ≤60 par/ml
Particle Count - 1.0 µm and greater ≤10 par/ml
For Microelectronic Use
Storage Note: Do not store near Ammonium Hydroxide. White crystals may form around the opening. The crystals can be removed with deionized water. This will not affect the quality of the product in the bottle.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR