Supelco® TraceCERT® Organic Reference Standard, Isobutyric acid
Proveedor: Merck
96499-100MG
96499-100MGEA 114 EUR
96499-100MG
Supelco® TraceCERT® Organic Reference Standard, Isobutyric acid
Normativas Estándares orgánicos
  • GHS signal: GHS02, GHS06, GHS05
Certificaciones: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Precaución: P210 - P233 - P280 - P301 + P312 - P303 + P361 + P353 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f2aa59a10af7f761aa5bf878fb29de3b005b1e1&vid=7f2aa59a10af7f761aa5bf878fb29de3b005b1e1&prid=web&clid=SAPDAM