Paladio 10% on carbon, seca for use as a catalyst Type 487

Proveedor: Thermo Fisher Scientific
Atención

A12012.04 A12012.06 A12012.09 A12012.14 A12012.18
A12012.04EA 117 EUR
A12012.04 A12012.06 A12012.09 A12012.14 A12012.18
Paladio 10% on carbon, seca for use as a catalyst Type 487
Palladium

CAS No.: 7440-05-3 Merck Reference: 14,6989 UN No.: 1325 MDL No.: MFCD00011167

Formula: Pd
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011167
Núm. CAS: 7440-05-3
EINECS: 231-115-6
UN: 1325
ADR: 4.1,III
Índice Merck: 14,6989

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR