D(+)-Biotina ≥98%

Proveedor: Thermo Fisher Scientific

Sinónimos: Vitamina B8, Vitamina B7, Vitamina H

A14207.06 A14207.03 A14207.09 A14207.14
A14207.06EA 256 EUR
A14207.06 A14207.03 A14207.09 A14207.14
D(+)-Biotina ≥98%
D(+)-Biotina

Purity: 98+% CAS No.: 58-85-5 Molecular Formula: C10H16N2O3S Formula Weight: 244.32 Storage Temperature: 2°C to 8°C Melting Point: 224-226° Beilstein Reference: 86838 Merck Reference: 14,1231 Optical Rotation: +91° (c=1 in 0.1N NaOH) MDL No.: MFCD00005541


Essential vitamin that is important for amino acid and energy metabolism

Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Pto. de fusión: 224…226 °C
Densidad: 1.41 g/cm³ (20 °C)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00005541
Núm. CAS: 58-85-5
EINECS: 200-399-3
Índice Merck: 14,1231

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR