Trimethylamine-N-oxide dihydrate ≥98%

Proveedor: Thermo Fisher Scientific
Atención

Sinónimos: Trimethylamine oxide dihydrate

A14916.14 A14916.06 A14916.22 A14916.36
A14916.14EA 47.3 EUR
A14916.14 A14916.06 A14916.22 A14916.36
Trimethylamine-N-oxide dihydrate ≥98%
Trimethylamine-N-oxide dihydrate

Purity: 98+% CAS No.: 62637-93-8 Molecular Formula: (CH3)3NO.2H2O Formula Weight: 111.14 (75.11anhy) Melting Point: 96-101° Beilstein Reference: 3612927 Merck Reference: 14,9710 MDL No.: MFCD00149077

Formula: C₃H₉NO·2H₂O
MW: 111.14 g/mol (75.11 anhy)
Pto. de fusión: 96…101 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00149077
Núm. CAS: 62637-93-8
EINECS: 214-675-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR