Ethylenediaminetetraacetic acid diammonium salt hydrate

Proveedor: Thermo Fisher Scientific

A15286.14 A15286.22 A15286.36
A15286.14EA 26 EUR
A15286.14 A15286.22 A15286.36
Ethylenediaminetetraacetic acid diammonium salt hydrate
Ethylenediaminetetraacetic acid diammonium salt hydrate

CAS No.: 20824-56-0 Molecular Formula: (NH4O2CCH2)2NCH2CH2N(CH2CO2H)2.xH2O Formula Weight: 326.31(anhy) Beilstein Reference: 7785165 MDL No.: MFCD00150463

Formula: C₁₀H₂₂N₄O₈·xH₂O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00150463
Núm. CAS: 20824-56-0
EINECS: 244-063-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR