Evans blue

Proveedor: Thermo Fisher Scientific
Peligro

A16774.09 A16774.18
A16774.09EA 64.2 EUR
A16774.09 A16774.18
Evans blue
Evans blue

CAS No.: 314-13-6 Molecular Formula: C34H24N6Na4O14S4 Formula Weight: 960.82 MDL No.: MFCD00004021


Applications: Direct Blue 53. Inhibits L-glutamate uptake, and AMPA and kainate receptor currents

Formula: C₃₄H₂₈N₆O₁₄S₄
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00004021
Núm. CAS: 314-13-6
EINECS: 206-242-5
Índice Merck: 14,3905

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR