L-Lysine methyl ester dihydrochloride 99%

Proveedor: Thermo Fisher Scientific

A18157.14 A18157.22 A18157.36
A18157.14EA 80.4 EUR
A18157.14 A18157.22 A18157.36
L-Lysine methyl ester dihydrochloride 99%
L-Lysine methyl ester dihydrochloride

Purity: 99% CAS No.: 26348-70-9 Molecular Formula: C7H16N2O2.2HCl Formula Weight: 233.14 Storage Temperature: 2°C to 8°C Melting Point: ca 200° dec. Beilstein Reference: 5302183 MDL No.: MFCD00039067

Formula: C₇H₁₆N₂O₂
Pto. de fusión: ∼ 200 °C (desc.)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00039067
Núm. CAS: 26348-70-9
EINECS: 247-625-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR