Trypan blue 60% Dye

Proveedor: Thermo Fisher Scientific
Peligro

Sinónimos: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

A18600.22 A18600.14 A18600.36
A18600.22EA 110 EUR
A18600.22 A18600.14 A18600.36
Trypan blue 60% Dye
Trypan blue

CAS No.: 72-57-1 Molecular Formula: C34H24N6Na4O14S4 Formula Weight: 960.82 Melting Point: 300° Merck Reference: 14,9792 MDL No.: MFCD00003969


Used in dye exclusion studies in cell culture

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003969
Núm. CAS: 72-57-1
EINECS: 200-786-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR