Fluoresceína 95

Proveedor: AAT BIOQUEST
Atención

Sinónimos: Ácido 2-(6-hidroxi-3-oxo-(3H)-xanten-9-il)benzoico

1
AATB1EA 120 EUR
AATB1
Fluoresceína 95
Fluoresceína

Fluorescein is a fluorophore widely used in microscopy, forensics and serology to detect latent blood stains.

Formula: C₂₀H₁₂O₅
MW: 332.31 g/mol
Número MDL: MFCD00005050
Núm. CAS: 2321-07-5
ADR: 5 II
Índice Merck: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b0e97bec9c9f44dd58e1cd0ca79d7e94c3ba82d&vid=a29b350acb5fd71b32a30baa408af54494e42d59&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR