Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 510)

Proveedor: AAT BIOQUEST
10033110 10033111
AATB10033110EA 371 EUR
AATB10033110 AATB10033111
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (mFluor™ Violet 510)
Anticuerpos
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: mFluor™ Violet 510
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR