Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (APC (Allophycocyanin)/iFluor® 750)

Proveedor: AAT BIOQUEST
100521G0 100521G1 100521G2
AATB100521G0EA 319 EUR
AATB100521G0 AATB100521G1 AATB100521G2
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (APC (Allophycocyanin)/iFluor® 750)
Anticuerpos
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: APC (Allophycocyanin)/iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR