Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet 450)

Proveedor: AAT BIOQUEST
103400Z0 103400Z1
AATB103400Z0EA 371 EUR
AATB103400Z0 AATB103400Z1
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet 450)
Anticuerpos
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Precaución: For research use only.

Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: mFluor™ Violet 450
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR