(Ac-KQL)2R110
Proveedor: AAT BIOQUEST
Atención
13465
AATB13465EA 152 EUR
AATB13465
(Ac-KQL)2R110
(Ac-KQL)2R110
The non-fluorescent R110 substrates generate the bright green fluorescent rhodamine 110 product that has Ex/Em = 494/521 nm, and can be easily detected with a FITC filter set.
  • Protease substrate
  • Bright signal
  • Multiple applications
ADR: 5,II