Amplite® Colorimetric L-Lactate Dehydrogenase (LDH) Assay Kit
Proveedor: AAT BIOQUEST
13813
AATB13813EA 460 EUR
AATB13813
Amplite® Colorimetric L-Lactate Dehydrogenase (LDH) Assay Kit
Ensayos Ensayos enzimáticos
Lactate dehydrogenase (LDH) is an oxidoreductase enzyme that catalyses the interconversion of pyruvate and lactate.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Precaución: For research use only.