Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Red 700
Proveedor: AAT BIOQUEST
16946
AATB16946EA 152 EUR
AATB16946
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Red 700
Proteínas y Péptidos
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
  • Optimized for flow cytometry
  • Useful for assay development
  • High affinity
  • Multiple applications
Precaución: For research use only.