Nuclear Orange™ DCS1
Proveedor: AAT BIOQUEST
17551
AATB17551EA 152 EUR
AATB17551
Nuclear Orange™ DCS1
Ensayos Ensayos celulares
Our Nuclear Orange™ DCS1 is a fluorogenic, DNA-selective and cell-impermeant dye for analyzing DNA content in dead, fixed or apoptotic cells.
  • Cell-Impermeable
  • Useful For Multicolor Imaging
  • Bright Fluorescence
5 mM DMSO Solution
Precaución: For research use only.