ReadiLink™ xtra Rapid Cy3 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
Proveedor: AAT BIOQUEST
1971
AATB1971EA 292 EUR
AATB1971
ReadiLink™ xtra Rapid Cy3 Antibody Labeling Kit *BSA-Compatible* (2 Labelings)
Sistemas de enlace y etiquetado de proteínas
ReadiLink™ xtra rapid antibody labelling kits require essentially only 2 simple mixing steps without a column purification needed.
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Precaución: For research use only.