Fura 2-AM ≥95%, Ultra Pure Grade

Proveedor: AAT BIOQUEST
Atención

21021 21023
AATB21021EA 230 EUR
AATB21021 AATB21023
Fura 2-AM ≥95%, Ultra Pure Grade
Fura 2-AM

Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.

Formula: C₄₄H₄₇N₃O₂₄
Número MDL: MFCD00036976
Núm. CAS: 108964-32-5
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR