MESG phosphate assay reagent
Proveedor: AAT BIOQUEST
21600-1MG
AATB21600-1MGEA 109 EUR
AATB21600-1MG
MESG phosphate assay reagent
MESG
In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.
  • Stable for 12 months
Quality tested in-house
Núm. CAS: 55727-10-1